ENA024296337 Enamine

2-{[(2,5-dimethylphenyl)sulfonyl]amino}-4-(methylthio)butanoic acid

  • MDL number MFCD05148153
  • Cas Number : 1008965-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10301

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-10301
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart