ENA024296340 Enamine

1-(4-fluorophenyl)-3-(3-methoxyphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06358033
  • Cas Number : 956198-12-2
  • Molecular Weight : 312.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10304
Availability details for
Quantity
  • Availability
  • Add to Cart