ENA024296348 Enamine

4-[(3,4-dihydro-2H-1,5-benzodioxepin-7-ylsulfonyl)amino]benzoic acid

  • MDL number MFCD06358068
  • Cas Number : 790272-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10312

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10312
Availability details for
Quantity
  • Availability
  • Add to Cart