ENA024296351 Enamine

4-amino-N-methyl-N-(1-methylpiperidin-4-yl)benzene-1-sulfonamide

  • MDL number MFCD06358021
  • Cas Number : 790272-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-137
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10315
Availability details for
Quantity
  • Availability
  • Add to Cart