ENA024296352 Enamine

4-{[(pentamethylphenyl)sulfonyl]amino}benzoic acid

  • MDL number MFCD06358069
  • Cas Number : 790272-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10316
Availability details for
Quantity
  • Availability
  • Add to Cart