ENA024296372 Enamine

2-[(aminocarbonyl)amino]-3-phenylpropanoic acid

  • MDL number MFCD00057654
  • Cas Number : 37534-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10336

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-10336
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart