ENA024296373 Enamine

N-(1-sec-butyl-1H-pyrazol-5-yl)-2-chloropropanamide

  • MDL number MFCD06360132
  • Cas Number : 956756-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10337

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10337
Availability details for
Quantity
  • Availability
  • Add to Cart