ENA024296386 Enamine

2-(4-isopropylphenyl)ethanamine

  • MDL number MFCD06212662
  • Cas Number : 84558-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10350
Availability details for
Quantity
  • Availability
  • Add to Cart