ENA024296400 Enamine

N-{[5-(2-chlorophenyl)-1,3,4-oxadiazol-2-yl]methyl}cyclopropanamine

  • MDL number MFCD06360136
  • Cas Number : 791079-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10364
Availability details for
Quantity
  • Availability
  • Add to Cart