ENA024296403 Enamine

2-(4-methylpiperidin-1-yl)-2-(thiophen-2-yl)ethan-1-amine

  • MDL number MFCD06360137
  • Cas Number : 791079-98-6
  • Molecular Weight : 224.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-methyl-1-piperidinyl)-2-(2-thienyl)ethylamine
Inchi Code 1S/C12H20N2S/c1-10-4-6-14(7-5-10)11(9-13)12-3-2-8-15-12/h2-3,8,10-11H,4-7,9,13H2,1H3
Inchi Key HJPGNUUFVKJORI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10367

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-10367
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart