ENA024296409 Enamine

7-methoxythieno[2,3-b]quinoline-2-carboxylic acid

  • MDL number MFCD02232019
  • Cas Number : 351358-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10373
Availability details for
Quantity
  • Availability
  • Add to Cart