ENA024296414 Enamine

1-(3,4-diethoxyphenyl)imidazolidine-2,4,5-trione

  • MDL number MFCD06345731
  • Cas Number : 775314-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10378

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10378
Availability details for
Quantity
  • Availability
  • Add to Cart