ENA024296417 Enamine

1-(3,4-dipropoxyphenyl)imidazolidine-2,4,5-trione

  • MDL number MFCD06345733
  • Cas Number : 775314-85-7
  • Molecular Weight : 306.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10381
Availability details for
Quantity
  • Availability
  • Add to Cart