ENA024296421 Enamine

4-[(3,4-dichlorobenzenesulfonamido)methyl]benzoic acid

  • MDL number MFCD06358012
  • Cas Number : 790272-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10385
Availability details for
Quantity
  • Availability
  • Add to Cart