ENA024296447 Enamine

2-bromo-5-(morpholine-4-sulfonyl)benzoic acid

  • MDL number MFCD00760906
  • Cas Number : 301156-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-129
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10411
Availability details for
Quantity
  • Availability
  • Add to Cart