ENA024296463 Enamine

2-[2-methoxy-4-(prop-2-en-1-yl)phenoxy]acetic acid

  • MDL number MFCD00017594
  • Cas Number : 6331-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10427
Availability details for
Quantity
  • Availability
  • Add to Cart