ENA024296469 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(3,4-dimethylphenyl)acetamide

  • MDL number MFCD06345781
  • Cas Number : 775315-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10433
Availability details for
Quantity
  • Availability
  • Add to Cart