ENA024296475 Enamine

4-[3-(trifluoromethyl)benzenesulfonamido]butanoic acid

  • MDL number MFCD01751667
  • Cas Number : 85844-93-5
  • Molecular Weight : 311.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10439
Availability details for
Quantity
  • Availability
  • Add to Cart