ENA024296488 Enamine

4-(1-benzofuran-2-yl)aniline

  • MDL number MFCD01078584
  • Cas Number : 782-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-174
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10452
Availability details for
Quantity
  • Availability
  • Add to Cart