ENA024296490 Enamine

2-[4-(3,4-dichlorobenzenesulfonamido)phenyl]acetic acid

  • MDL number MFCD06335572
  • Cas Number : 749896-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10454
Availability details for
Quantity
  • Availability
  • Add to Cart