ENA024296494 Enamine

2-chloro-N-(4-chloro-1,1-dioxo-1lambda6-thiolan-3-yl)acetamide

  • MDL number MFCD05694679
  • Cas Number : 757221-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10458
Availability details for
Quantity
  • Availability
  • Add to Cart