ENA024296497 Enamine

2-(3-chloro-4-fluorobenzenesulfonamido)-4-(methylsulfanyl)butanoic acid

  • MDL number MFCD03619000
  • Cas Number : 1009235-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10461
Availability details for
Quantity
  • Availability
  • Add to Cart