ENA024296501 Enamine

5-amino-2-{[(5-tert-butyl-2-methylphenyl)sulfonyl]amino}-5-oxopentanoic acid

  • MDL number MFCD06334883
  • Cas Number : 1009483-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10465

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10465
Availability details for
Quantity
  • Availability
  • Add to Cart