ENA024296504 Enamine

4-(chloromethyl)-1,3-diphenyl-1H-pyrazole

  • MDL number MFCD02714800
  • Cas Number : 55432-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10468
Availability details for
Quantity
  • Availability
  • Add to Cart