ENA024296505 Enamine

2-chloro-N-[1-(2-naphthyl)ethyl]acetamide

  • MDL number MFCD06358026
  • Cas Number : 790272-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10469

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10469
Availability details for
Quantity
  • Availability
  • Add to Cart