ENA024296510 Enamine

1-[(5-sulfanyl-1,3,4-oxadiazol-2-yl)methyl]pyrrolidin-2-one

  • MDL number MFCD06358045
  • Cas Number : 160758-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10474
Availability details for
Quantity
  • Availability
  • Add to Cart