ENA040898933 Enamine

2-(1H-1,3-benzodiazol-2-yl)-N-butylacetamide

  • MDL number MFCD06655052
  • Cas Number : 91600-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10491
Availability details for
Quantity
  • Availability
  • Add to Cart