ENA040898944 Enamine

3-(4-ethylphenyl)-1-(2-methylphenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06655063
  • Cas Number : 956763-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10528

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10528
Availability details for
Quantity
  • Availability
  • Add to Cart