ENA040898950 Enamine

[2-amino-1-(2-chlorophenyl)ethyl]diethylamine

  • MDL number MFCD06655069
  • Cas Number : 851169-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10542
Availability details for
Quantity
  • Availability
  • Add to Cart