ENA040898956 Enamine

(2E)-3-methyl-2-[2-(2-methylphenyl)-2-oxoethylidene]-1,3-thiazolidin-4-one

  • MDL number MFCD06655075
  • Cas Number : 855715-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10574
Availability details for
Quantity
  • Availability
  • Add to Cart