ENA040898960 Enamine

3-[6-(chloromethyl)-3-methyl-4-oxo-1H,4H,7H-pyrazolo[3,4-b]pyridin-1-yl]-1lambda6-thiolane-1,1-dione

  • MDL number MFCD06655078
  • Cas Number : 855715-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10580
Availability details for
Quantity
  • Availability
  • Add to Cart