ENA040898970 Enamine

2-mercapto-3-(4-methylpyridin-2-yl)quinazolin-4(3H)-one

  • MDL number MFCD06364475
  • Cas Number : 793716-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10605

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10605
Availability details for
Quantity
  • Availability
  • Add to Cart