ENA040898971 Enamine

8-chloro-1-(chloromethyl)-4-(4-ethoxyphenyl)-4H,5H-[1,2,4]triazolo[4,3-a]quinazolin-5-one

  • MDL number MFCD06655085
  • Cas Number : 855715-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10606
Availability details for
Quantity
  • Availability
  • Add to Cart