ENA040898980 Enamine

5-(4-fluorophenyl)-2-mercapto-3-(2-phenylethyl)thieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD06660499
  • Cas Number : 851169-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10629
Availability details for
Quantity
  • Availability
  • Add to Cart