ENA040898983 Enamine

2-(4-methyl-2,5-dioxo-4-phenylimidazolidin-1-yl)acetic acid

  • MDL number MFCD06655093
  • Cas Number : 726-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10639
Availability details for
Quantity
  • Availability
  • Add to Cart