ENA040899003 Enamine

1-[2,4-bis(propan-2-yl)phenyl]-2-chloroethan-1-one

  • MDL number MFCD06655109
  • Cas Number : 164165-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10682
Availability details for
Quantity
  • Availability
  • Add to Cart