ENA040899007 Enamine

3-{[(3-bromophenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD06351000
  • Cas Number : 785792-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10686

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10686
Availability details for
Quantity
  • Availability
  • Add to Cart