ENA040899015 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(4-ethylphenyl)acetamide

  • MDL number MFCD06655114
  • Cas Number : 855715-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10699
Availability details for
Quantity
  • Availability
  • Add to Cart