ENA040899017 Enamine

4-oxo-3-(2-phenylethyl)-2-sulfanyl-3,4-dihydroquinazoline-7-carboxylic acid

  • MDL number MFCD03030057
  • Cas Number : 422271-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10701
Availability details for
Quantity
  • Availability
  • Add to Cart