ENA040899018 Enamine

methyl 2-mercapto-3-(3-methoxypropyl)-4-oxo-3,4-dihydroquinazoline-7-carboxylate

  • MDL number MFCD05024778
  • Cas Number : 451465-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10702
Availability details for
Quantity
  • Availability
  • Add to Cart