ENA040899020 Enamine

2-mercapto-3-(pyridin-3-ylmethyl)quinazolin-4(3H)-one

  • MDL number MFCD00104425
  • Cas Number : 422274-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10704

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10704
Availability details for
Quantity
  • Availability
  • Add to Cart