ENA040899024 Enamine

4-[(3-methylpiperidin-1-yl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06655121
  • Cas Number : 855715-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10708
Availability details for
Quantity
  • Availability
  • Add to Cart