ENA040899031 Enamine

2-(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)acetohydrazide

  • MDL number MFCD06655123
  • Cas Number : 121565-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-153
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10716
Availability details for
Quantity
  • Availability
  • Add to Cart