ENA040899039 Enamine

2-(chloromethyl)-1-ethyl-5-(piperidin-1-ylsulfonyl)-1H-benzimidazole

  • MDL number MFCD06351004
  • Cas Number : 785792-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234...236
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10733
Availability details for
Quantity
  • Availability
  • Add to Cart