ENA040899042 Enamine

2-(chloromethyl)-1-ethyl-5-(morpholin-4-ylsulfonyl)-1H-benzimidazole

  • MDL number MFCD06350981
  • Cas Number : 785792-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10736
Availability details for
Quantity
  • Availability
  • Add to Cart