ENA040899049 Enamine

2-chloro-N-{spiro[1,3-dioxaindane-2,1'-cyclopentan]-6-yl}acetamide

  • MDL number MFCD06655128
  • Cas Number : 855715-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10746
Availability details for
Quantity
  • Availability
  • Add to Cart