ENA040899055 Enamine

N-[3-(chloroacetyl)-2,5-dimethyl-1H-pyrrol-1-yl]benzamide

  • MDL number MFCD06353498
  • Cas Number : 781626-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10754

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10754
Availability details for
Quantity
  • Availability
  • Add to Cart