ENA040899058 Enamine

4-tert-butyl-2,6-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD02018257
  • Cas Number : 70823-04-0
  • Molecular Weight : 260.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-114
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10758

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-10758
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart