ENA040899059 Enamine

2-(4-methylpiperazin-1-yl)-2-(thiophen-2-yl)ethan-1-amine

  • MDL number MFCD06655134
  • Cas Number : 857041-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-59
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10759
Availability details for
Quantity
  • Availability
  • Add to Cart