ENA040899061 Enamine

N-(2-amino-1-phenylethyl)-N,N-diethylamine

  • MDL number MFCD06655136
  • Cas Number : 31788-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10761

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-10761
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart